missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher LaboClean FM

Product Code. 15459493
Change view
Click to view available options
Volume (Metric):
5 L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15459493 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15459493 Supplier Dr.Weigert™ Supplier No. 406633
  • ONLINE EXCLUSIVE PRICE
    134.00 €
    94.96 € / 5 L
    Save 39.04 €
    29% Off
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Neodisher LaboClean FM - Alkaline intensive detergent, liquid concentrate. For automated cleaning of laboratory glassware and other utensils for laboratories in the food industry, the cosmetics and pharmaceutical industry, the mineral oil industry and in other industries.

  • Alkaline intensive detergent, liquid concentrate
  • Neodisher LaboClean FM removes of oil, fat, resin, paraffin and organic dyes
  • Free of surfactants
  • Free of phosphates, silicates and oxidising agents
  • Good emulsifying and dispersing action.
  • Especially suitable for the removal of oil, fat, resins and paraffin.
  • For professional use only
  • Dosage between 2-5 mL/L

Specifications

Form Liquid
pH 12.0 to 12.4
Sterility Non-sterile
Composition <5 % amphoteric surfactants 5 - 15 % polycarboxylates
Concentration 2 to 5 mL/L
Container Type Canister
Density 1.2 g/cm3 (20°C)
Description 220 x 240 x 140 mm
Filtered Filtered
Viscosity <50 mPa.s (20°C)
Volume (Metric) 5 L
Show More Show Less
compliance-icons
Product Identifier
  • neodisher LaboClean FM
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2-[carboxymethyl(2-hydroxyethyl)amino]acetic acid, sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.