missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher LM 3

Product Code. 10589071
Change view
Click to view available options
Volume (Metric):
10 L
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10589071 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10589071 Supplier Dr.Weigert™ Supplier No. 440030
  • ONLINE EXCLUSIVE PRICE
    175.00 €
    165.87 € / 10 L
    Save 9.13 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Neodisher LM 3 - Midly alkaline cleaning agent liquid without phosphate. Manual cleaning of laboratory glassware in immersion baths in Medical, dental practices, industrial and Biological laboratories. Can be used in immersion and ultra sonic baths.

  • Mildly alkaline liquid cleaning agent
  • Neodisher LM2 is used for manual cleaning laboratory instruments and anesthesia utensils, while protecting them against corrosion
  • Offers high protection for laboratory glass, stainless steel, instrument steel, pure aluminum, aluminum-magnesium alloys, chromium-plated devices, and customary plastics
  • Not create foam
  • Suited for use in special washers for e.g. viscosimeters or pipettes
  • Free of phosphates
  • Effective against blood, egg and food source residues
  • Can be used with all water hardness levels
  • Suggested dose is 5-30 mL/L
  • Specifications

    Product Type Neodisher LM 3
    Form Liquid
    pH 12.3 to 13.3
    Sterility Non-sterile
    Composition <5 % amphoteric surfactants
    Concentration 2 to 200 mL/L
    Density 1.1 g/cm3
    Description 310 x 230 x 180 mm
    Filtered Filtered
    Volume (Metric) 10 L
    compliance-icons
    Product Identifier
    • neodisher LM 3
    Signal Word
    • Danger
    Hazard Category
    • Substances and mixtures corrosive to metals Category 1
    • Skin corrosion/irritation Category 1 B
    • Serious eye damage/eye irritation Category 1
    Hazard Statement
    • H290-May be corrosive to metals.
    • H314-Causes severe skin burns and eye damage.
    • EUH208-Contains N-(2-hydroxyethyl)-N-[2-[(1-oxooctyl)amino]ethyl-ß-alanine. May produce an allergic reaction.
    Precautionary Statement
    • P280-Wear protective gloves/protective clothing/eye protection/face protection.
    • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
    • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    • P310-Immediately call a POISON CENTER/doctor/.
    Supplemental information
    • MIXTURE LIST-Contain: sodium cumenesulfonate, N-(2-hydroxyethyl)-N-[2-[(1-oxooctyl)amino]ethyl-ß-alanine, potassium hydrox
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.