missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher MA

Product Code. 11784644
Change view
Click to view available options
pH:
11.5 to 12.1
Unit Size:
10kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Unit Size
11784644 11.5 to 12.1 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11784644 Supplier Dr.Weigert™ Supplier No. 410776
  • ONLINE EXCLUSIVE PRICE
    136.00 €
    114.64 € / 10kg
    Save 21.36 €
    15% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Neodisher MA - Powder detergent free of oxidative additives used for machine cleaning of surgical instruments, anesthesia instruments, surgical footwear and baby bottles in hospitals, as well as laboratory glassware.

  • Mildly alkaline powder detergent
  • Neodisher MA is used for cleaning laboratory instruments and anesthesia utensils, while protecting them against corrosion
  • Offers high protection for laboratory glass, stainless steel, instrument steel, pure aluminum, aluminum-magnesium alloys, chromium-plated devices, and customary plastics
  • Effective against blood, egg and food source residues
  • Reduces foam
  • Can be used with all water hardness levels
  • Suggested dose is 1-3 g/L
  • Titration factor of 0.44
  • Specifications

    Product Type Neodisher MA
    Form Powder
    pH 11.5 to 12.1
    Sterility Non-sterile
    Appearance White
    Certifications/Compliance Conforms to the requirements of the legislation for medical devices according to European directive 93/42/EC
    Composition <5 % non-ionic surfactants > 30 % phosphates
    Concentration 1 to 4 g/L
    Container Type Bucket
    Content And Storage Store away from moisture, with lid closed, for up to 2 years.
    Density 915 - 965 g/L
    Description 280 x 270 mm
    For Use With (Application) Machine Cleaning of Laboratory Instruments and Anesthesia Utensils Made of Glass, Stainless / Instrument Steel, Aluminum, Aluminum-Magnesium Alloy, Chromium-Plated and Plastic.
    Viscosity 915 - 965 g/L
    Volume (Metric) 10 kg
    Show More Show Less
    compliance-icons compliance-icons
    Product Identifier
    • neodisher MA
    Signal Word
    • Danger
    Hazard Category
    • Skin corrosion/irritation Category 1 B
    • Serious eye damage/eye irritation Category 1
    • Specific target organ toxicity Category 3
    Hazard Statement
    • H314-Causes severe skin burns and eye damage.
    • H335-May cause respiratory irritation.
    Precautionary Statement
    • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
    • P280-Wear protective gloves/protective clothing/eye protection/face protection.
    • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
    • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    • P310-Immediately call a POISON CENTER/doctor/.
    Supplemental information
    • MIXTURE LIST-Contain: fatty alcohol, ethoxylated, disodium metasilicate pentahydrate
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.