missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher N

Product Code. 10313801
Change view
Click to view available options
Dimensions (L x W x H):
250 x 100 x 60 mm
220 x 240 x 140 mm
250 x 190 x 140 mm
390 x 290 x 240 mm
Container Type:
Bottle
Canister
Unit Size:
25kg
5 L
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Container Type Unit Size
10313801 220 x 240 x 140 mm Canister 5 L
11785773 250 x 100 x 60 mm Bottle Each
15377833 220 x 240 x 140 mm Canister 5 L
11728393 250 x 190 x 140 mm Canister 5 L
10331302 390 x 290 x 240 mm Canister 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10313801 Supplier Dr.Weigert™ Supplier No. 420133
  • ONLINE EXCLUSIVE PRICE
    209.10 €
    157.17 € / 5 L
    Save 51.93 €
    24% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Neodisher N - Acidic detergent and neutralising agent liquid. Neutralizes by adding to the first rinse after use of an alkaline detergent. Automated acidic cleaning of animal cages.

  • Based on phosphoric acid
  • neodisher N is used for neutralizing alkaline residues in washer disinfection
  • Available in either bottles or canisters
  • Has very good rinsing properties
  • Suitable for use with: surgical instruments, anesthesia items, containers, surgical footwear, baby bottles, and other medical devices, as well as laboratory glassware
  • Free of surfactantsDosage of between 2-4 mL/L
  • Titration factor: 0.16

Specifications

Product Type Neodisher N
Form Liquid
pH 2.2 to 1.8
Sterility Non-sterile
Composition Phosphoric acid, citric acid
Concentration 1 to 4 mL/L
Container Type Canister
Dimensions (L x W x H) 220 x 240 x 140 mm
Filtered Filtered
For Use With (Application) Neutralizing Any Carried Over Alkaline Residues on Laboratory Glassware or for Acid Pre-Cleaning Resp
Volume (Metric) 5 L
Show More Show Less
compliance-icons
Product Identifier
  • neodisher N
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: phosphoric acid, citric acid anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.