missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher PM5

Product Code. 11873242
Change view
Click to view available options
Volume (Metric):
3 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11873242 3 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11873242 Supplier Dr.Weigert™ Supplier No. 440281
  • ONLINE EXCLUSIVE PRICE
    85.00 €
    72.85 € / Pack of 1
    Save 12.15 €
    14% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Neodisher PM 5 - Midly Alkaline cleaning agent in powder form for use in immersion baths. For manual cleaning of laboratory in hospital, medical and biological laboratories. Removes food residues, inorganic, organic and gynaecological residues such as lubricants.

  • Midly alkaline detergent, Powder
  • neodisher LaboClean PM 5 is used for manual cleaning laboratory glassware and pre-cleaning of reprocessable medical devices, particulary for gyneacological instruments
  • oxygen-based bleaching agents, phosphates and non-ionic surfactants and anionic surfactants
  • Suitable for all types of laboratories utensils light metals equipment and stainless steel
  • Very easily rinsed off
  • Suitable for all water hardness
  • Dosage of 5-20 g/L
  • action

Specifications

Product Type Neodisher PM5 Cleaner Manual
Form Powder
pH 11.2 to 11.8
Sterility Non-sterile
Composition 5% anionic surfactants, non-ionic surfactants, oxygen-based bleaching agents > 30% phophates.
Concentration 5 to 20 g/L
Density 1,100 - 1,200 g/L
Description 220 x 140 mm
Volume (Metric) 3 kg
compliance-icons
Product Identifier
  • neodisher PM 5
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: trisodium phosphate,sodium carbonate peroxyhydrate, sodiumcarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.