missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher TS

Product Code. 10247800
Change view
Click to view available options
Volume (Metric):
10 L
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10247800 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10247800 Supplier Dr.Weigert™ Supplier No. 310930
  • ONLINE EXCLUSIVE PRICE
    128.00 €
    106.53 € / 10 L
    Save 21.47 €
    16% Off
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Neodisher TS - Rinse aid liquid. Neutralizes alkaline residues during automated reprocessing. Designed for dishes, food containers and trolleys.

  • Rinse aid With non-ionic surfactants and phosphonates
  • neodisher TS is used in specially designed for dishwasher
  • Suitable also in washer disinfection in Medical
  • formula highly concentrated
  • Acidic formulation
  • Neutralizes alkaline residues
  • Dosage of between 0.1-0.8 mL/L
  • Neutralizes alkaline residues

Specifications

Product Type Neodisher TS
Form Liquid
pH 4.2 to 3.5
Sterility Non-sterile
Composition 15 - 30 % nonionic surfactants, < 5 % phosphonates, also preservatives (methylchloroisothiazolinone)
Concentration 0.1 to 0.8 mL/L
Density 1.1 g/cm3 (20°C)
Description 310 x 230 x 180 mm
Filtered Filtered
Volume (Metric) 10 L
compliance-icons
Product Identifier
  • neodisher TS
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: citric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.