missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher LaboClean UW

Product Code. 11743733
Change view
Click to view available options
Volume (Metric):
10 kg
Unit Size:
10kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11743733 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11743733 Supplier Dr.Weigert™ Supplier No. 410976
  • ONLINE EXCLUSIVE PRICE
    236.00 €
    197.18 € / 10kg
    Save 38.82 €
    16% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Neodisher LaboClean UW - Designed for machine cleaning of laboratory glass and utensils in water laboratories, serological institutes and in the phosphate industry. Neodisher LaboClean UW is a detergent powder free of phosphates for use in sensitive analyses where traces of phosphorus would interfere.

  • Mildly alkaline detergent powder
  • Neodisher laboclean UW is used to with material protection and good cleanability for inorganic and organic contaminants
  • Composed of non-ionic surfactants and oxygen-bases bleaching agents
  • Free of phosphates
  • Dosage of between 3-5 g/L (depending on the degree of contamination)

Specifications

Product Type Neodisher UW
Form Powder
pH 11.7 to 11.9 (3 to 5 g/L, water, 20°C)
Sterility Non-sterile
Appearance White
Composition Non-ionic surfactants, bleach based on oxygen
Concentration 3 to 5 g/L
Container Type Bucket
Content And Storage Store at 0-25, away from moisture, for up to 2 years.
Density 970 g/L
Description 280 x 270 mm
For Use With (Application) Machine Cleaning of Laboratory Glass and Utensils.
Volume (Metric) 10 kg
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • neodisher LaboClean UW
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: fatty alcohol, ethoxylated, disodium metasilicate pentahydrate, sodium carbonateperoxyhydrate, sodi
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.