missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) nitride, 99.9% (REO)

Product Code. 11339737
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11339737

Brand: Thermo Scientific Alfa Aesar 042091.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25764-11-8 | NNd | 158.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25764-11-8
Molecular Formula NNd
Molecular Weight (g/mol) 158.249
MDL Number MFCD00049990
InChI Key OVMJQLNJCSIJCH-UHFFFAOYSA-N
Synonym neodymium nitride, neodymium nitride ndn, neodymium iii nitride, neodymiumylidyneamine
PubChem CID 117628
IUPAC Name azanylidyneneodymium
SMILES N#[Nd]

Specifications

Physical Form Powder
Quantity 10 g
Formula Weight 158.25
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Neodymium(III) nitride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.