missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III)-oxide, 99.9%, (trace metal basis)

Product Code. 10606982 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10606982 100 g Glass bottle 100g
10686802 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10606982 Supplier Thermo Scientific Acros Supplier No. 211581000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1313-97-9 | Nd2O3 | 336.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-97-9
Molecular Formula Nd2O3
Molecular Weight (g/mol) 336.48
MDL Number MFCD00011134
InChI Key PLDDOISOJJCEMH-UHFFFAOYSA-N
IUPAC Name dineodymium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Nd+3].[Nd+3]

Specifications

Melting Point 2270.0°C
Color Blue
CAS Min % 99.85
CAS Max % 100.0
Physical Form Powder
Quantity 100 g
Packaging Glass bottle
Specific Gravity 7.24
Assay Percent Range 99.90%
Density 7.2400g/mL
Linear Formula ND2O3
Merck Index 14, 6450
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in acids
Formula Weight 336.47
Percent Purity 99.9%
Grade Trace Metal Basis
Chemical Name or Material Neodymium(III)-oxide, 99.9%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.