missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) oxide, REacton™, 99.997% (REO), Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 013837.22

Quantity 100g

 View more versions of this product

Product Code. 15401057

  • 497.00 € / 100g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

1313-97-9
MFCD00011134

Specifications

1313-97-9
MFCD00011134
Insoluble in water. Soluble in acids.
99.997%
7.24 g/mL
Neodymium(III) oxide
100g
14,6450
336.48
Reacton™
(REO)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.