missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Neodymium(III) titanate

Product Code. 11355079
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11355079 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11355079 Supplier Alfa Aesar™ Supplier No. A17366.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 12058-94-5
Molecular Formula Nd2Ti2O7
MDL Number MFCD00192342
Synonym Neodymium titanium oxide

Specifications

Quantity 5 g
Formula Weight 496.21
Odor Odorless
Chemical Name or Material Neodymium(III) titanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.