missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Neodymium(III) titanate

Product Code. p-7023620
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 11355079

Brand: Alfa Aesar™ A17366.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 12058-94-5
Molecular Formula Nd2Ti2O7
MDL Number MFCD00192342
Synonym Neodymium titanium oxide

Specifications

Quantity 5 g
Formula Weight 496.21
Odor Odorless
Chemical Name or Material Neodymium(III) titanate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.