missing translation for 'onlineSavingsMsg'
Learn More

Neomycin C Hexaacetate, TRC

Product Code. 30482259
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482259 1 mg 1mg
30474889 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30482259 Supplier Toronto Research Chemicals Supplier No. TRCN3899801MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Neomycin C Hexaacetate
Purity Grade Notes HPLC
Molecular Formula C23H46N6O13 • 6(CH3COOH)
Quantity 1 mg
Synonym O-2,6-Diamino-2,6-dideoxy-α-D-glucopyranosyl-(1→3)-O-β-D-ribofuran osyl-(1→5)-O-[2,6-diamino-2,6-dideoxy-α-D-glucopyranosyl-(1→4)]-2-deoxy-D-streptamine Hexaacetate, Dekamycin II Hexaacetate
SMILES N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CN)O[C@@H]1O[C@H]2[C@@H](O)[C@H](O[C@H]3[C@H](O[C@@H]4[C@H](N)[C@@H](O)[C@H](O)[C@@H](CN)O4)[C@@H](N)C[C@@H](N)[C@@H]3O)O[C@@H]2CO
Molecular Weight (g/mol) 614.6466005
Percent Purity >95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.