missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Neon™ NxT Electroporation System 10-μL Kit with 1-Channel Tubes

Product Code. 18075331
Change view
Click to view available options
No. of Reactions:
25 x 2 Reactions
96 x 2 Reactions
Unit Size:
Each
Pack of 25
Pack of 96
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
18075331 25 x 2 Reactions Pack of 25
17829444 25 x 2 Reactions Each
17839444 96 x 2 Reactions Each
18035341 96 x 2 Reactions Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18075331 Supplier Invitrogen™ Supplier No. N1025
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Neon NxT Electroporation System 10-μL Kit is designed specifically for use with the Neon NxT Electroporation System.

The Neon NxT Electroporation System 10-μL Kit with 1-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System (Cat. Nos. NEON1S, NEON1SK, NEON18S, NEON18SK). Use this kit for transfection volumes of 10 μL, containing 5 x 104–2 x 105 adherent cells or 1 x 105–5 x 105 suspension cells. Cells that have been transfected using the included 10-μL Neon NxT tips are ready to be washed and plated into a multi-well culture dish.

The Neon NxT Electroporation System 10-μL Kit with 1-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System (Cat. Nos. NEON1S, NEON1SK, NEON18S, NEON18SK). Use this kit for transfection volumes of 10 μL, containing 5 x 104–2 x 105 adherent cells or 1 x 105–5 x 105 suspension cells. Cells that have been transfected using the included 10-μL Neon NxT tips are ready to be washed and plated into a multi-well culture dish.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1 mL Resuspension R Buffer; store at 4°C
• 1 mL Resuspension T Buffer; store at 4°C
• 1 mL Resuspension Genome Editing Buffer; store at 4°C
• 50 mL Electrolytic E10 Buffer; store at 4°C
• 25 reaction tips
• 8 electroporation 1-channel tubes
Cell Type Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format Electroporation Pipette Tip
Transfection Technique Electroporation
For Use With (Equipment) Neon™ NxT Electroporation System
For Use With (Application) Transfection
No. of Reactions 25 x 2 Reactions
Product Type Pipette Tip
Quantity 25 x 2 reactions
Volume (Metric) Sample 10 μL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.