Learn More
Invitrogen™ Neon™ NxT Electroporation System 8-Channel Pipette Station
Product Code. 30245339
Change view
Click to view available options
Quantity:
1 pipette station
Unit Size:
Each
Product Code. Quantity Unit Size
30245339 1 pipette station Each
1 options
Product Code. 30245339 Supplier Invitrogen™ Supplier No. NEON8PS
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Neon NxT Electroporation System 8-Channel Pipette Station is designed specifically for use with the Neon NxT Electroporation System.
The Neon NxT Electroporation System 8-Channel Pipette Station is designed specifically for use with the Neon NxT Electroporation System (Cat. No. NEON18S). Each Neon NxT Electroporation System with 1-Channel and 8-Channel Pipettes (Cat. No. NEON18S and NEON18SK) comes with an 8-channel pipette station. Those looking to upgrade their Neon NxT Electroporation System with 1-Channel Pipette (Cat. No. NEON1S, NEON1SK) should pursue the Neon NxT Electroporation System 8-Channel Upgrade Package (Cat. No. NEON8 and NEON8U), which includes the Neon NxT Electroporation System 8-Channel Pipette, 8-channel pipette station, and 8-channel software activation. This pipette station is offered separately for those needing an additional 8-channel pipette station or replacement for their existing pipette station.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Transfection
For Use With (Equipment) Neon™ NxT Electroporation System
Quantity 1 pipette station