Learn More
Invitrogen™ Neon™ NxT Electroporation System 8-Channel Upgrade Package
Description
The Neon NxT Electroporation System 8-Channel Upgrade Package is available for the Neon NxT Electroporation System with 1-Channel Pipette (Cat. No. NEON1S and NEON1SK). The upgrade package includes the Neon NxT 8-channel pipette, 8-channel pipette station, and 8-channel software activation.
The Neon NxT Electroporation System 8-Channel Pipette enables the electroporation of 96 samples in less than 15-minutes while allowing for full flexibility over each reaction. Due to the 8-channel tube design, each tip is separated in its own chamber and isolated from the other tips, allowing for different electroporation parameters, resuspension buffer, delivery payload, or even cell type. Additionally, the Neon NxT 8-channel pipette utilizes the same Neon NxT tips and buffers as the 1-channel pipette.
Specifications
Specifications
| Compatible Tips | Neon™ NxT |
| Number of Channels | 8 |
| Volume (Metric) | 10 μL, 100 μL |
| For Use With (Application) | Transfection |
| For Use With (Equipment) | Neon™ NxT Electroporation System (Cat. No. NEON1S, NEON1SK) |
| Includes | Neon™ NxT 8-channel pipette, 8-channel pipette station, 8-channel software activation, and quick reference guide |
| Quantity | 1 package |