missing translation for 'onlineSavingsMsg'
Learn More

Neopentyl alcohol, 99%

Product Code. 10058581 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10058581 10 g Glass bottle 10g
10520712 50 g Glass bottle 50g
10697304 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10058581 Supplier Thermo Scientific Acros Supplier No. 128250100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-84-3 | C5H12O | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-84-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004682
InChI Key KPSSIOMAKSHJJG-UHFFFAOYSA-N
Synonym neopentyl alcohol, 2,2-dimethyl-1-propanol, 2,2-dimethylpropanol, tert-butylcarbinol, 1-propanol, 2,2-dimethyl, tert-butyl carbinol, neopentanol, neopentylalcohol, neoamyl alcohol, 2,2-dimethylpropyl alcohol
PubChem CID 6404
IUPAC Name 2,2-dimethylpropan-1-ol
SMILES CC(C)(C)CO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 52°C to 53°C
Color Colorless
Density 0.8100g/mL
Boiling Point 113°C to 114°C
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CCH2OH
Quantity 10 g
Beilstein 01, 406
Merck Index 15, 6542
Specific Gravity 0.81
Solubility Information Solubility in water: 3.5% (25 c)
Formula Weight 88.15
Percent Purity 99%
Physical Form Waxy Crystalline Solid
Chemical Name or Material Neopentyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • Neopentyl alcohol
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.