missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Neprilysin (MME) ELISA Kit

Product Code. 15415073
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15415073 96 Tests 1 set
17145509 5 x 96 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415073 Supplier Invitrogen™ Supplier No. EHMME

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Human Neprilysin (MME) ELISA quantitates Hu MME in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu MME. Principle of the method The Human MME solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

CD10 (Common Acute Lymphocytic Leukemia Antigen, CALLA), is a cell surface enzyme with neutral metalloendopeptidase activity which inactivates a variety of biologically active peptides. CD10 is expressed on the cells of lymphoblastic, Burkitt's, and follicular germinal center lymphomas, immature B cells with in adult bone marrow and on cells from patients with chronic myelocytic leukemia (CML). CD10 is also present on breast myoepithelial cells, bile canaliculi, fibroblasts, with especially high expression on the brush border of kidney and gut epithelial cells. CD10 is a neutral endopeptidase that cleaves peptides at the amino side of hydrophobic residues and inactivates several peptide hormones including glucagon, enkephalins, substance P, neurotensin, oxytocin, and bradykinin. Further, CD10 is a 100 kDa type II transmembrane glycoprotein that exists in a single copy of greater than 45 kb. The 5' untranslated region of the CD10 gene is alternatively spliced, resulting in four separate mRNA transcripts and the coding region is not affected by alternative splicing. Diseases associated with CD10 dysfunction include spinocerebellar ataxia 43 and Charcot-Marie tooth Disease.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08473
Assay Range 0.41 to 100 ng/mL
Assay Sensitivity 0.4 ng/mL
Calibration Range 0.4 ng/mL to 100 ng/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Product Type ELISA
Protein Family Cytokines & Receptors
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: human ADAM-9, Angiopoietin 2, APRIL, BMP-2, BMP-9, C5a, Cathepsin L, CD200, CD97, Chemerin, DcR3, FABP2, FAP, FGF-19, Galectin-3, HGF R, IFN-gammalpha, IGF-2, IGF-2 R, IL-1R6, IL-24, IL-33, Kallikrein 14, Legumain, LOX-1, MBL, Notch-1, NOV, Osteoactivin, PD-1, PGRP-5, Serpin A4, sFRP-3, Thrombomodulin, TLR2, TRAIL R1, Transferrin, WIF-1 ACE-2, Albumin, AMICA, Angiopoietin 4, BAFF, CA19-9, CD163 , Clusterin, CRTAM, CXCL14, Cystatin C, Decorin , Dkk-3, DLL1, Fetuin A, aFGF, FOLR1, Furin, GASP-1, GASP-2, GCSF R, HAI-2, IL-17 B R, IL-27, LAG-3 , LDL R, Pepsinogen I, RANK, RBP4, SOST, Syndecan-1, TACI, TFPI, TSP-1, TRAIL R2 , TRANCE, Troponin I, uPA, VE-Cadherin, WISP-1
Sufficient For 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 4311
Gene Symbol CALLA,CD10,CMT2T,NEP,SCA43,SFE
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Cell Cycle & Proliferation
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Synonym CALLA, CD10, NEP, SFE
Target CD10
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.