missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Boc-L-lysine methyl ester hydrochloride, 98%

Product Code. 15420569
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420569 5 g 5g
15410569 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15420569 Supplier Thermo Scientific Alfa Aesar Supplier No. H63727.06
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2389-48-2 | C12H25ClN2O4 | 296.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in methanol.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2389-48-2
Molecular Formula C12H25ClN2O4
Molecular Weight (g/mol) 296.79
MDL Number MFCD00076959
InChI Key NANRHOPPXCBHGI-MVRPGTNWNA-N
Synonym h-lys boc-ome.hcl, h-lys boc-ome hcl, h-lys boc-ome hydrochloride, lys boc-ome.hcl, s-methyl 2-amino-6-tert-butoxycarbonyl amino hexanoate hydrochloride, methyl 2s-2-amino-6-tert-butoxycarbonyl amino hexanoate hydrochloride, pubchem18976, h-lys boc-ome?cl, lys boc-ome. hcl, h-lys boc-ome??hcl
PubChem CID 16218818
IUPAC Name methyl (2S)-2-amino-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate;hydrochloride
SMILES Cl.COC(=O)[C@@H](N)CCCCNC(=O)OC(C)(C)C

Specifications

Beilstein 5173577
Solubility Information Soluble in methanol.
Quantity 5 g
Formula Weight 296.79
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Nepsilon-Boc-L-lysine methyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.