missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neratinib, MedChemExpress
Description
Neratinib (HKI-272) is an orally available, irreversible, highly selective HER2 and EGFR inhibitor with IC50s of 59 nM and 92 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | Neratinib |
| CAS | 698387-09-6 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C30H29ClN6O3 |
| Quantity | 200 mg |
| Synonym | HKI-272 |
| Solubility Information | DMSO : 13.33 mg/mL (23.93 mM; Need ultrasonic) |
| SMILES | ClC1=C(OCC2=NC=CC=C2)C=CC(NC3=C(C#N)C=NC4=CC(OCC)=C(NC(/C=C/CN(C)C)=O)C=C43)=C1 |
| Molecular Weight (g/mol) | 557.04 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?