missing translation for 'onlineSavingsMsg'
Learn More

Neratinib

Product Code. 16514444 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
16514444 250 mg 250mg
16504444 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 16514444

Brand: Thermo Scientific Acros 464702500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 698387-09-6 | C30H29ClN6O3 | 557.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 698387-09-6
Molecular Formula C30H29ClN6O3
Molecular Weight (g/mol) 557.05
InChI Key JWNPDZNEKVCWMY-UHFFFAOYSA-N
IUPAC Name N-[4-({3-chloro-4-[(pyridin-2-yl)methoxy]phenyl}amino)-3-cyano-7-ethoxyquinolin-6-yl]-4-(dimethylamino)but-2-enamide
SMILES CCOC1=C(NC(=O)C=CCN(C)C)C=C2C(NC3=CC(Cl)=C(OCC4=CC=CC=N4)C=C3)=C(C=NC2=C1)C#N

Specifications

Quantity 250 mg
Formula Weight 557.05
Chemical Name or Material Neratinib
compliance-icons compliance-icons
Product Identifier
  • Neratinib, Thermo Scientific Chemicals
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Neratinib

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.