Learn More
Thermo Scientific™ NERL™ High Purity Water
Improve control in critical diagnostic procedures by using Thermo Scientific™ NERL™ High Purity Water in place of deionized or distilled water.
Brand:  Thermo Scientific™ 9805
Quantity 5 gal.
 View more versions of this product
Product Code. 15312208
  • 93.00 € / Pack of 1
Estimated Shipment: 23-02-2027
Add to basket
Alert:
  • For in vitro diagnostic and laboratory use only
  • Not for infusion, injection or irrigation
Description
Description
Improve control in critical diagnostic procedures by using Thermo Scientific™ NERL™ High Purity Water in place of deionized or distilled water. This product meets CAP/CLSI specifications for Clinical Laboratory Reagent Grade Water (CLRW) at time of manufacture.
Typically prepared at 18 megohm/cm specific resistance using reverse osmosis, mixed bed deionization, activated carbon filtration and final filtration to 0.2 micron, NERL high purity water is essentially free from organic and inorganic particulate and soluble contaminants.
Contains no preservatives
Ordering Information:
Certificate of analysis available for each case
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 to 7
NERL
5 gal.