missing translation for 'onlineSavingsMsg'
Learn More

Neryl acetate, 98%

Product Code. 11429546
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429546 100 g 100g
11419546 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429546 Supplier Thermo Scientific Alfa Aesar Supplier No. A19282.22
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 141-12-8 | C12H20O2 | 196.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-12-8
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00063205
InChI Key HIGQPQRQIQDZMP-FLIBITNWSA-N
Synonym neryl acetate, nerol acetate, neryl ethanoate, z-3,7-dimethylocta-2,6-dien-1-yl acetate, nerol acetate 6ci, unii-of82iju18h, cis-3,7-dimethyl-2,6-octadien-1-yl acetate, cis-geranyl acetate, cis-3,7-dimethyl-2,6-octadien-1-ol acetate, of82iju18h
PubChem CID 1549025
IUPAC Name [(2Z)-3,7-dimethylocta-2,6-dienyl] acetate
SMILES CC(=CCCC(=CCOC(=O)C)C)C

Specifications

Density 0.91
Boiling Point 133°C to 135°C (25 mmHg)
Flash Point 98°C (208°F)
Odor Pleasant
Linear Formula (CH3)2C=CHCH2CH2(CH3)C=CHCH2CO2CH3
Refractive Index 1.46
Quantity 100 g
Beilstein 1722814
Formula Weight 196.29
Percent Purity 98%
Chemical Name or Material Neryl acetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.