missing translation for 'onlineSavingsMsg'
Learn More

Neuraminidase, Clostridium perfringens

Product Code. 15475529
Change view
Click to view available options
Quantity:
10 mg
Packaging:
Plastic bottle
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15475529 10 mg Plastic bottle 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15475529 Supplier Thermo Scientific Alfa Aesar Supplier No. J63259.MA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9001-67-6

Neuraminidase is used as a hydrolytic enzyme that removes sialic acid from mucoproteins, used in a study to assess binding with human T lymphocytes in sheep pretreated with neuraminidase. It has also been used in a study to investigate the effect of bile salts on the action of hydrolysis by neuraminidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 9001-67-6
Packaging Plastic bottle
MDL Number MFCD00131711
Quantity 10 mg
Synonym Acyl-neuraminyl hydrolase,EC 3.2.1.18
Solubility Information Soluble in water or 10mM phosphate buffer,pH 6.0
Physical Form Lyophilized Powder
Chemical Name or Material Neuraminidase

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.