missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies NeuroCult™ Neuronal Plating Medium

Product Code. 17198281
Encompass
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17198281 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17198281 Supplier STEMCELL Technologies Supplier No. 05713
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

NeuroCult™ Neuronal Plating Medium is a serum-free neuronal basal medium

  • NeuroCult™ Neuronal Plating Medium is a serum-free neuronal basal medium
  • It is designed for use with BrainPhys™ Neuronal Medium (Catalog #05790) for the plating and culture of primary tissue-derived neurons

Specifications

Cell Type Neurons
For Use With (Application) Cell Culture; Maintenance
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.