missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies NeuroCult™ NS-A Basal Medium (Human)

Product Code. 17148291
Encompass
Change view
Click to view available options
Quantity:
450 mL
Unit Size:
450mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17148291 450 mL 450mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17148291 Supplier STEMCELL Technologies Supplier No. 05750
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Basal medium for expansion of human neural stem and progenitor cells

  • NeuroCult™ NS-A Basal Medium (Human) is a standardized basal medium for the expansion of human neural stem cells and brain tumor stem cells, in the neurosphere or adherent monolayer culture system
  • This basal medium is a component of the NeuroCult™ NS-A Proliferation Kit (Human; Catalog #05751) and the NeuroCult™ NS-A Differentiation Kit (Human; Catalog #05752)

Specifications

Cell Type Brain Tumor Stem Cells, Neural Stem and Progenitor Cells
For Use With (Application) Cell Culture; Colony Assay; Differentiation; Expansion; Functional Assay; Spheroid Culture
Quantity 450 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.