missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies NeuroCult™-XF Proliferation Medium

Product Code. 17178291
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17178291 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17178291 Supplier STEMCELL Technologies Supplier No. 05761
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Xeno-free medium for expansion of human neural stem and progenitor cells

  • NeuroCult™-XF Proliferation Medium is a standardized, xeno-free, serum-free medium for the culture of human neural stem and progenitor cells from normal tissues or tumor samples, in the neurosphere or adherent monolayer system
  • When supplemented with appropriate cytokines, NeuroCult™-XF Proliferation Medium is optimized to maintain human neural stem cells in culture for extended periods of time without the loss of their self-renewal, proliferation or differentiation potential
  • NOTE: Addition of Carrier-Free rh EGF (Catalog #78006) and rh bFGF (Catalog #78003) is required
  • Heparin (Catalog #07980) can also be added, however, the heparin solution contains non-human animal-derived components and can be omitted if a complete xeno-free system is desired

Specifications

Cell Type Brain Tumor Stem Cells, Neural Stem and Progenitor Cells
For Use With (Application) Cell Culture, Colony Assay, Expansion, Functional Assay, Spheroid Culture
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.