missing translation for 'onlineSavingsMsg'
Learn More

Decon™ Neutracon

Product Code. 10355650
Change view
Click to view available options
Volume (Metric):
1000 mL
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10355650 1000 mL 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10355650 Supplier Decon™ Supplier No. NEU1
  • ONLINE EXCLUSIVE PRICE
    17.18 €
    14.75 € / 1 L
    Save 2.43 €
    14% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Decon Neutracon, Quantity: 5000mL, Amber, Format: Liquid, pH Range: 7.5, Viscosity: 55.0cps at 23 deg.C

Biodegradable, phosphate, enzyme and chlorine bleach free, near-neutral liquid surface-active agent. Completely soluble in water and will not deposit calcium and magnesium or precipitate iron. Solutions are completely stable in air and do not absorb CO&superscript_2;. May be used in conjunction with water soluble solvents. A 13% solution is sufficient for the majority of cleaning applications. Heating the solution, or use in an ultrasonic cleaning bath will greatly reduce the cleaning time. Specifically formulated for cleaning/decontamination of non-ferrous metals, 'soft' silica glass, polymer surfaces and other materials which may be corroded, etched, degraded, discolored or weakened by acidic or alkaline cleaning agents.

Specifications

Form Liquid
pH 7.5
Appearance Amber
Viscosity 55.0cps at 23°C
Volume (Metric) 1000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.