missing translation for 'onlineSavingsMsg'
Learn More

Decon™ Neutracon

Product Code. p-8014318
Click to view available options
Volume (Metric):
1000 mL
Unit Size:
1 L
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 10355650

Brand: Decon™ NEU1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Decon Neutracon, Quantity: 5000mL, Amber, Format: Liquid, pH Range: 7.5, Viscosity: 55.0cps at 23 deg.C

Biodegradable, phosphate, enzyme and chlorine bleach free, near-neutral liquid surface-active agent. Completely soluble in water and will not deposit calcium and magnesium or precipitate iron. Solutions are completely stable in air and do not absorb CO&superscript_2;. May be used in conjunction with water soluble solvents. A 13% solution is sufficient for the majority of cleaning applications. Heating the solution, or use in an ultrasonic cleaning bath will greatly reduce the cleaning time. Specifically formulated for cleaning/decontamination of non-ferrous metals, 'soft' silica glass, polymer surfaces and other materials which may be corroded, etched, degraded, discolored or weakened by acidic or alkaline cleaning agents.

Specifications

Form Liquid
pH 7.5
Appearance Amber
Viscosity 55.0cps at 23°C
Volume (Metric) 1000 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.