Learn More
Syngene™ Neutral Field Screen Set for use with G:BOX Systems
Product Code. 12814038 Shop All Syngene Products
Change view
Click to view available options
For Use With (Equipment):
G:BOX systems
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
12814038 G:BOX systems Each
1 options
Product Code. 12814038 Supplier Syngene™ Supplier No. GXNFS
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Syngene™ Neutral Field Screen Set for use with G:BOX Systems that includes a blue screen and a frosted screen.
  • Corrects for any uneven Illumination by acquiring a neutral-field reference image with the same ntensity illumination as the experiment
  • For use with Epi-UV lighting and transilluminators
Specifications
For Use With (Equipment) G:BOX systems
Includes 1 blue screen and 1 frosted screen
Dimensions (L x W x H) 335 x 330 x 15 mm