missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Neutral Red

Product Code. 15475319
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15475319 1 g 1g
15485319 5 g 5g
15495319 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15475319 Supplier Thermo Scientific Chemicals Supplier No. J63054.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nuclear counterstain useful in pH range 6.8 to 8.0 | CAS: 553-24-2 | C15H17ClN4 | 288.779 g/mol

Nuclear counterstain useful in pH range 6.8 - 8.0

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-24-2
Molecular Formula C15H17ClN4
Molecular Weight (g/mol) 288.779
MDL Number MFCD00012651
InChI Key PGSADBUBUOPOJS-UHFFFAOYSA-N
Synonym Basic Red 5; C.I. 50040
PubChem CID 11105
ChEBI CHEBI:86370
IUPAC Name 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride
SMILES CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl

Specifications

Melting Point 290°C (decomposition)
Beilstein 3918740
Merck Index 14,6488
Color Black to Green
Quantity 1 g
Physical Form Powder
Formula Weight 288.78
Chemical Name or Material Neutral Red

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.