missing translation for 'onlineSavingsMsg'
Learn More

Melford Neutral Red

Product Code. 30018573
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018573 5 g 5g
30018572 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018573 Supplier Melford Supplier No. N224005.0
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Biological dye used to stain nuclei red and in testing viability of cells to incorporate and bind Neutral Red.

Biological dye used to stain nuclei red and in testing viability of cells to incorporate and bind Neutral Red. Dye Content: >90%

Specifications

Chemical Name or Material Neutral Red
CAS 553-24-2
Molecular Formula C15H16N4-HCl
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.