missing translation for 'onlineSavingsMsg'
Learn More

Melford Neutral Red

Product Code. p-200087210
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 30018573

Brand: Melford N224005.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biological dye used to stain nuclei red and in testing viability of cells to incorporate and bind Neutral Red.

Biological dye used to stain nuclei red and in testing viability of cells to incorporate and bind Neutral Red. Dye Content: >90%

Specifications

Chemical Name or Material Neutral Red
CAS 553-24-2
Molecular Formula C15H16N4-HCl
Quantity 5 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.