missing translation for 'onlineSavingsMsg'
Learn More

Neutral Red (Technical Grade), TRC

Product Code. 30478292
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478292 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478292 Supplier Toronto Research Chemicals Supplier No. TRCN391013100G
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Neutral Red (Technical Grade)
CAS 553-24-2
Molecular Formula C15H16N4 .HCl
Quantity 100 g
Synonym C.I. Basic Red 5','Monohydrochloride, Toluylene Red, 3-Amino-7-dimethylamino-2-methylphenazine Hydrochloride, C.I. 50040, C.I. Basic Red 5, Neutral Red, Neutral Red W, Nuclear Fast Red (basic dye), N8,N8,3-Trimethyl-2,8-phenazinediamine Monohydrochloride
InChI Formula InChI=1S/C15H16N4.ClH/c1-9-6-13-15(8-11(9)16)18-14-7-10(19(2)3)4-5-12(14)17-13;/h4-8H,16H2,1-3H3;1H
SMILES Cl.CN(C)c1ccc2nc3cc(C)c(N)cc3nc2c1
IUPAC Name 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride
Molecular Weight (g/mol) 288.77
Formula Weight 288.1142
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.