missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Neutravidin Protein, DyLight™ 800

Product Code. 10280054
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10280054 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10280054 Supplier Invitrogen™ Supplier No. 22853
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Protein

22853 contains 1 mL of protein in pale green colored liquid at a concentration of 1 mg/mL.

Avidin is a glycoprotein found in the egg white and tissues of birds, reptiles and amphibia. The biotin-binding protein contains four identical subunits having a combined mass of 67,000-68,000 daltons. Removing the glycosylation from avidin yields NeutrAvidin Protein with a mass of 60,000 daltons. Carbohydrate-based lectin-binding is reduced to undetectable levels, yet biotin-binding affinity is retained. NeutrAvidin Protein offers the advantages of a neutral pI to minimize nonspecific adsorption, along with lysine residues that remain available for derivatization or other customized conjugation. NeutrAvidin Protein yields the lowest nonspecific binding among the known biotin binding proteins. The specific activity for biotin-binding is approximately 14 μg/mg of protein, which is near the theoretical maximum activity.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL, 1mg/mL
For Use With (Application) ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.2
Name Neutravidin
Conjugate DyLight 800
Cross Reactivity Tag
pH Range 7.2
Purification Method Purified
Quantity 1 mL
Storage Requirements 4°C
Regulatory Status RUO
pH 7.2
Common Name NeutrAvidin
Product Type Protein, Neutravidin
Content And Storage 4°C, Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
Form Liquid
Purity or Quality Grade purified
Host Species N/A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.