missing translation for 'onlineSavingsMsg'
Learn More

Nevirapine, MedChemExpress

Product Code. 16494048
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494048 10 mg 10mg
16450407 50 mg 50mg
16435036 100 mg 100mg
16430996 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494048 Supplier Medchem Express Supplier No. HY10570
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nevirapine is a non-nucleoside inhibitor of HIV-1 reverse transcriptase used to treat and prevent HIV/AIDS; with a Ki of 270 μM.

Nevirapine is a non-nucleoside inhibitor of HIV-1 reverse transcriptase used to treat and prevent HIV/AIDS

Specifications

Chemical Name or Material Nevirapine
CAS 129618-40-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H14N4O
Quantity 10 mg
Synonym BI-RG 587 NSC 641530 NVP
Solubility Information DMSO : 14.29 mg/mL (53.66 mM; Need ultrasonic)
SMILES O=C1C2=C(N=CC=C2)N(C3CC3)C4=NC=CC(C)=C4N1
Molecular Weight (g/mol) 266.3
Formula Weight 266.3
Percent Purity 99.01%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.