missing translation for 'onlineSavingsMsg'
Learn More

Nevirapine, TRC

Product Code. 30010805
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010805 50 mg 50mg
30007722 25 mg 25mg
17951895 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010805 Supplier Toronto Research Chemicals Supplier No. TRCN39127550MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129618-40-2
Molecular Formula C15 H14 N4 O
Molecular Weight (g/mol) 266.3
Synonym Nevirapine Anhydrous, 6H-Dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one, 11-cyclopropyl-5,11-dihydro-4-methyl- (9CI), BI-RG 587, NSC 641530, Nevarapine, Nevirapine, Viramune
IUPAC Name 11-cyclopropyl-4-methyl-5H-dipyrido[2,3-b:3',2'-e][1,4]diazepin-6-one
SMILES Cc1ccnc2N(C3CC3)c4ncccc4C(=O)Nc12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H14N4O/c1-9-6-8-17-14-12(9)18-15(20)11-3-2-7-16-13(11)19(14)10-4-5-10/h2-3,6-8,10H,4-5H2,1H3,(H,18,20)
Formula Weight 266.1168
Chemical Name or Material Nevirapine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.