missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals New Coccine, pure

Product Code. 10055943 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10055943 100 g Glass bottle 100g
10583361 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10055943 Supplier Thermo Scientific Chemicals Supplier No. 190231000
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 2611-82-7 | C20H11N2Na3O10S3 | 604.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2611-82-7
Molecular Formula C20H11N2Na3O10S3
Molecular Weight (g/mol) 604.46
MDL Number MFCD00004084
InChI Key IVNZBWNBYXERPK-DZGBHZPSSA-K
Synonym Acid Red 18, C.I. 16255, Cochineal Red A
PubChem CID 131851859
IUPAC Name (8Z)-7-oxo-8-[(4-sulfonaphthalen-1-yl)hydrazinylidene]naphthalene-1,3-disulfonic acid;sodium
SMILES [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=C4C=CC=CC4=C(C=C3)S([O-])(=O)=O)C(=O)C=C2)C(=C1)S([O-])(=O)=O

Specifications

Dye Content 75% min.
Beilstein 16, I, 306
Infrared Spectrum Authentic
Formula Weight 604.46
Loss on Drying 10% max. (135°C, 6 hrs)
Grade Extra Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Bordeaux to Brown
Quantity 100 g
Physical Form Powder
Chemical Name or Material New Coccine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.