missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Newborn Bovine Serum

Product Code. 17347479 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17347479 100 mL 100mL
11405025 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17347479 Supplier MP Biomedicals™ Supplier No. 092912149
  • ONLINE EXCLUSIVE PRICE
    36.55 €
    34.65 € / 100mL
    Save 1.90 €
    5% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in cell culture to support the maturation of oocytes, to study the function of tubular epithelial cells, and to develop primary cultures of cells such as visceral adipocytes.

Specifications

Color Golden Brown
Content And Storage -20°C
Product Type Newborn Calf Bovine Serum
Quantity 100 mL
Sterilization Method 0.2 μm filtration
Source Bovine Neonate
Endotoxin Concentration 30EU/mL
For Use With (Application) Cell culture and organ preservation applications
Hemoglobin Concentration ≤27mg/dL
Lyophilized No
Mycoplasma Detected No
Osmolality 265 to 323mOsm/kg
pH 7.1 to 8.2
Protein Content 4.4 to 7%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.