missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress NFAT Inhibitor-1

Product Code. 16479498
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479498 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16479498 Supplier MedChemExpress Supplier No. HYP1026
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress NFAT Inhibitor (VIVIT peptide) is a cell-permeable peptide inhibitor of nuclear factor of activated Tcells (NFAT) that selectively inhibits calcineurin-mediated dephosphorylation of NFAT.

Specifications

Purity 99.86%
CAS 249537-73-3
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) COVID-19-immunoregulation
Molecular Weight (g/mol) 1685.94 g/mol
Product Type Peptides
Quantity 10 mg
Shipping Condition Room temperature
Solubility H2O : 100 mg/mL (59.31 mM∣Need ultrasonic)
Synonym VIVIT peptide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.