missing translation for 'onlineSavingsMsg'
Learn More

NFAT Transcription Factor Regulator-1, MedChemExpress

Product Code. p-200100375
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485526

Brand: Medchem Express HY112778

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NFAT Transcription Factor Regulator-1 is an IL-2 synthesis inhibitor with an IC50 of 182 nM.

NFAT Transcription Factor Regulator-1 is an IL-2 synthesis inhibitor with an IC50 of 182 nM.

Specifications

Chemical Name or Material NFAT Transcription Factor Regulator-1
CAS 245747-71-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H10F6N4O2
Quantity 100 mg
Solubility Information DMSO : ≥ 150 mg/mL (360.33 mM)
SMILES O=C(C1=C(F)C=NC=C1)NC2=CC=C(N3N=C(C(F)(F)F)C=C3OC(F)F)C=C2
Molecular Weight (g/mol) 416.28
Formula Weight 416.28
Percent Purity 98.98%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.