Learn More
Abcam NFkB p50 Transcription Factor Activity Assay Kit
Product Code. 32114452 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 x 96 Tests
Unit Size:
96 tests
Product Code. Quantity Unit Size
32114452 1 x 96 Tests 96 tests
1 options
Product Code. 32114452 Supplier Abcam Supplier No. ab3242021x96Tests
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Measure NFkB p105 / p50 activity in 1h 30m in Cell/Tissue Extracts with NFkB p50 Transcription Factor Activity Assay Kit AB324202. Sensitivity >0.1875 μg. Use with Pre-coated microplate (12 x 8 well strips).
Measure NFkB p105 / p50 activity in 1h 30m in Cell/Tissue Extracts with NFkB p50 Transcription Factor Activity Assay Kit AB324202. Sensitivity >0.1875 μg. Use with Pre-coated microplate (12 x 8 well strips).
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1 x 96 Tests