Learn More
LNI Swissgas NG CASTORE BASIC 200
Product Code. 17862309 Shop All LNI Swissgas Products
Change view
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
Product Code. Voltage unitSize
17862309 100/115/220/230 V Each
1 options
Product Code. 17862309 Supplier LNI Swissgas Supplier No. 6920.52.200
Nitrogen generator with no dryer.
  • Needs source of dry and oil free air.
Specifications
Voltage 100/115/220/230 V
Type Nitrogen Gas Generator