Learn More
LNI Swissgas NG CASTORE BASIC 45.40
Product Code. 17822029 Shop All LNI Swissgas Products
Change view
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
Product Code. Voltage Unit Size
17822029 100/115/220/230 V Each
1 options
Product Code. 17822029 Supplier LNI Swissgas Supplier No. 6920.52.046
  • ONLINE EXCLUSIVE PRICE
    8525.00 €
    7391.26 € / Pack of 1
    Save 1133.74 €
    13% Off
View all promotions
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Nitrogen generator with air outlet and no dryer.
  • Needs source of dry and oil free air.
Specifications
Voltage 100/115/220/230 V
Type Nitrogen Gas Generator