missing translation for 'onlineSavingsMsg'
Learn More

NG,NG’-Dimethy-L-arginine Di(p-hydroxyazobenzene-p’-sulfonate) Salt, TRC

Product Code. 30011567
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011567 5 mg 5mg
30133355 10 mg 10mg
30009984 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011567 Supplier Toronto Research Chemicals Supplier No. TRCD4635805MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102783-24-4
Molecular Formula 2 C12 H10 N2 O4 S . C8 H18 N4 O2
Molecular Weight (g/mol) 758.82
Synonym L-Ornithine, N5-[(dimethylamino)iminomethyl]-, bis[4-[(4-hydroxyphenyl)azo]benzenesulfonate] (9CI), Benzenesulfonic acid, 4-[(4-hydroxyphenyl)azo]-, compd. with N5-[(dimethylamino)iminomethyl]-L-ornithine (2:1) (9CI), NG,NG-Dimethyl-L-arginine di(p-hydroxyazobenzene-p'-sulfonate) salt, N5-[(Dimethylamino)iminomethyl]-L-ornithine bis[4-[(4-hydroxyphenyl)azo]benzenesulfonate] salt, (2S)-2-Amino-5-[[amino(dimethylamino)methylidene]amino]pentanoic acid 4-[(4-hydroxyphenyl)azo]benzenesulfonic acid (1:2), (S)-2-Amino-5-(3,3-dimethylguanidino)pentanoic acid di(p-hydroxyazobenzene-p'-sulfonate) salt, Asymmetric dimethylarginine di(p-hydroxyazobenzene-p'-sulfonate) salt, NG,NG-Dimethyl-L-arginine bis[4-[(4-hydroxyphenyl)azo]benzenesulfonate] salt
IUPAC Name (2S)-2-amino-5-[(N,N-dimethylcarbamimidoyl)amino]pentanoic acid;4-[(E)-(4-hydroxyphenyl)diazenyl]benzenesulfonic acid
SMILES CN(C)C(=N)NCCC[C@H](N)C(=O)O.Oc1ccc(cc1)N=Nc2ccc(cc2)S(=O)(=O)O.Oc3ccc(cc3)N=Nc4ccc(cc4)S(=O)(=O)O

Specifications

InChI Formula InChI=1S/2C12H10N2O4S.C8H18N4O2/c2*15-11-5-1-9(2-6-11)13-14-10-3-7-12(8-4-10)19(16,17)18;1-12(2)8(10)11-5-3-4-6(9)7(13)14/h2*1-8,15H,(H,16,17,18);6H,3-5,9H2,1-2H3,(H2,10,11)(H,13,14)/b2*14-13+;/t;;6-/m..0/s1
Quantity 5 mg
Formula Weight 758.2152
Shipping Condition Room Temperature
Chemical Name or Material NG,NG'-Dimethy-L-arginine Di(p-hydroxyazobenzene-p'-sulfonate) Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.