Learn More
LNI Swissgas NG SIRIO 1500 NC
Product Code. 17852309 Shop All LNI Swissgas Products
Change view
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
Product Code. Voltage unitSize
17852309 100/115/220/230 V Each
1 options
Product Code. 17852309 Supplier LNI Swissgas Supplier No. 6913.02.150
Nitrogen generator ultra high purity.
  • Needs source of air
Specifications
Voltage 100/115/220/230 V
Type Ultra High Purity Nitrogen Gas Generator