Learn More
LNI Swissgas NGA CASTORE XL iQ 40.40
Product Code. 17852139 Shop All LNI Swissgas Products
Change view
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
Product Code. Voltage unitSize
17852139 100/115/220/230 V Each
1 options
Product Code. 17852139 Supplier LNI Swissgas Supplier No. 6920.70.41
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
Nitrogen generator.
  • With integrated compressor.
Specifications
Voltage 100/115/220/230 V
Type Nitrogen Gas Generator