missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NHI 2

Description
NHI 2 is an LDHA inhibitor; reduces lactate production in HeLa cells. Inhibits cell growth of LPC006 and PANC-1 cells in vitro. Enhances the antiproliferative effects of Gemcitabine (Cat. No. 3259) on LPC006 and PANC-1 cells. Also inhibits ferroptosis.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | NHI 2 |
| Purity | >98% |
| Molecular Weight (g/mol) | 335.28 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?