missing translation for 'onlineSavingsMsg'
Learn More

NHP Compensation Set, Clone: L200, SP34-2, SK1, BD

Product Code. 15809548
Change view
Click to view available options
Quantity:
20 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15809548 20 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15809548 Supplier BD Biosciences Supplier No. 558640
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Conjugate FITC/APC/PE
Purification Method Affinity Purified
Quantity 20 Tests
Isotype IgG1 κ, IgG1 λ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.