Learn More
Descrizione
NHS Mag Sepharose are magnetic beads designed for pull-down techniques enabling rapid capture and enrichment of selected proteins based on affinity. An antibody or other protein with desired affinity is covalently coupled to the medium via primary amines for the purpose of capture of target proteins of interest.
- Visible and dense Sepharose based magnetic beads with N-hydroxysuccinimide (NHS) ligand, easy to spot and collect bound target protein
- Non-adherent beads eliminate smearing effects and aggregate formation. Can be used without detergents
- Simple capture of target protein in small or large sample volumes (low microliter to high milliliter scale)
- Optimized capacity for enrichment/pull-down of target proteins - only low amounts of bound capture molecule
- Flexible protocols with elution conditions optimized for downstream applications such as electrophoresis and MS
Matrix: Paramagnetic, spherical, highly cross-linked agarose particles
Ligand: N-hydroxysuccinimide
Ligand density: 8-14μmol/mL gel
Particle size: 37-100μm
Working temperature: Room temperature
Storage solution: 20% 2-propanol
Storage temperature: 4°C to 8°C
Specifica
Specifica
| Bead Type | NHS Mag Sepharose |
| Binding Capacity | 8 to 14μM/mL |
| Ligand Concentration | 8 to 14μM/mL |
| Matrix | Paramagnetic spherical, highly cross linked agarose particles |
| Particle Size | 37 to 100 μm |
| Quantity | 4 x 500 μL |
| Content And Storage | 4°C to 8°C |
| For Use With (Application) | Designed for covalent coupling of any Protein or biomolecule containing a free Amine group and is used for various types of pull down experiments, for biomarker research |
| Ligand | N-hydroxysuccinimide |
Sicurezza e movimentazione
- NHS Mag Sepharose, 4 x 500 ul
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- UNKNOWN AQUATIC-Contains 15% of components with unknown hazards to the aquatic environment.
- UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.